SY3408585616 Synthonix Corporation

Methyl 2-(tert-butoxycarbonylamino)thiazole-5-carboxylate

  • MDL number MFCD11655929
  • Cas Number : 745078-03-9
  • Molecular Weight : 258.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/10072.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/10072.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart