SY3408585617 Synthonix Corporation

[3-(1-Naphthyl)phenyl]boronic acid

  • MDL number MFCD11045054
  • Cas Number : 881913-20-8
  • Molecular Weight : 248.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/6929.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/6929.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart