SY3412111633 Synthonix Corporation

1-(Azetidin-3-yl)-3-methylazetidin-3-ol di-trifluoroacetate

  • MDL number MFCD26961269
  • Cas Number : 1651840-83-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/6327.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/6327.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart