SY3424062588 Synthonix Corporation

(1-Methyl-3-oxo-cyclobutyl)carbamic acid tert-butyl ester

  • MDL number MFCD27986945
  • Cas Number : 1523617-99-3
  • Molecular Weight : 199.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl (1-methyl-3-oxocyclobutyl)carbamate
Inchi Code 1S/C10H17NO3/c1-9(2,3)14-8(13)11-10(4)5-7(12)6-10/h5-6H2,1-4H3,(H,11,13)
Inchi Key DPLRVTIHVYBKPN-UHFFFAOYSA-N

Description

Purity 97.00%
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/10144.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/10144.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart