SY3424062591 Synthonix Corporation

6-Aminotetrahydro-2,2-dimethyl-4H-cyclopenta-1,3-dioxol-4-ol

  • MDL number No MDL found
  • Cas Number : 155899-66-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/10118.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/10118.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart