SY3424062599 Synthonix Corporation

endo-8-Boc-3-hydroxy-8-azabicyclo[3.2.1]octane

  • MDL number No MDL found
  • Cas Number : 143557-91-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/10150.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/10150.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart