SY3424062618 Synthonix Corporation

1-Methyl-3-phenyl-1H-pyrazole-5-carbaldehyde

  • MDL number No MDL found
  • Cas Number : 864068-96-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/9920.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/9920.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart