SY3425143501 Synthonix Corporation

2,5-Dibromopyridine-3,4-diamine

  • Cas Number : 221241-11-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/8891.html

Safety Information

MSDS https://synthonix.com/8891.html
Availability details for
Quantity
  • Availability
  • Add to Cart