SY3425143552 Synthonix Corporation

N-Methyl-N-(piperidin-4-yl)methanesulfonamide

  • Cas Number : 70724-74-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/9001.html

Safety Information

MSDS https://synthonix.com/9001.html
Availability details for
Quantity
  • Availability
  • Add to Cart