SY3428346881 Synthonix Corporation

trans-1-Boc-3-cyano-4-hydroxypyrrolidine

  • MDL number No MDL found
  • Cas Number : 215922-85-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/11639.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/11639.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart