SY3428346945 Synthonix Corporation

4-(Boc-aminomethyl)-4-fluoropiperidine

  • MDL number No MDL found
  • Cas Number : 871022-62-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/10613.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/10613.html
hstatements H302 , H315 , H318 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart