SY3428347133 Synthonix Corporation

trans-3-(Boc-amino)-4-hydroxypiperidine

  • MDL number MFCD22373823
  • Cas Number : 859854-68-5
  • Molecular Weight : 216.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/11608.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/11608.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart