SY3428347213 Synthonix Corporation

2-Aza-spiro[3.4]octane hemioxalate

  • MDL number MFCD27988100
  • Cas Number : 1523617-94-8
  • Molecular Weight : 312.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/12064.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/12064.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart