SY3428347307 Synthonix Corporation

3-Benzyl-7-oxo-3-aza-bicyclo[3.3.1nonan-9-one

  • MDL number No MDL found
  • Cas Number : 77716-01-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/10692.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/10692.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart