SY3428347352 Synthonix Corporation

1H-Benzimidazole-7-carboxaldehyde

  • MDL number No MDL found
  • Cas Number : 144876-36-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/10980.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/10980.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart