SY3428347386 Synthonix Corporation

8-Bromo-7-methoxy-1,6-naphthyridin-4(1H)-one

  • MDL number MFCD21642042
  • Cas Number : 952138-17-9
  • Molecular Weight : 255.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/11302.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/11302.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart