SY3428347580 Synthonix Corporation

4-Chlorothieno[2,3-d]pyrimidine-6-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 86825-96-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/11228.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/11228.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart