SY3428347691 Synthonix Corporation

2,4-Dichloro-5-methyl-5H-pyrrolo[3,2-d]pyrimidine

  • MDL number No MDL found
  • Cas Number : 129872-81-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/10687.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/10687.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart