SY3428347791 Synthonix Corporation

(S)-1,4-Diazabicyclo[4.3.0]nonane hydrobromide

  • MDL number No MDL found
  • Cas Number : 150208-84-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/11717.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/11717.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart