SY3428347859 Synthonix Corporation

Ethyl trans-3-hydroxycyclobutanecarboxylate

  • MDL number No MDL found
  • Cas Number : 160351-88-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/10943.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/10943.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart