SY3428348011 Synthonix Corporation

Ethyl 5-bromopyrazolo[1,5-a]pyridine-2-carboxylate

  • MDL number No MDL found
  • Cas Number : 1363381-49-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin USA
Product Link https://synthonix.com/11337.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/11337.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart