SY3428348039 Synthonix Corporation

N-Hydroxy-5-methyl-1H-Indole-3-carboximidamide

  • MDL number No MDL found
  • Cas Number : 889943-61-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/10698.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/10698.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart