SY3428348065 Synthonix Corporation

2-(1H-1,2,4-triazol-3-yl)acetic acid

  • MDL number MFCD21909522
  • Cas Number : 155049-60-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/11339.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/11339.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart