SY3428348075 Synthonix Corporation

2-Hydroxymethyl-5-methylpyrimidine

  • MDL number No MDL found
  • Cas Number : 90905-61-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/11473.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/11473.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart