SY3428348078 Synthonix Corporation

tert-butyl N-[(3R,5S)-5-(hydroxymethyl)pyrrolidin-3-yl]carbamate

  • MDL number No MDL found
  • Cas Number : 1264243-41-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/11522.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/11522.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart