SY3428348081 Synthonix Corporation

6-Boc-6H-octahydro-pyrrolo[3,4-b]pyridine

  • MDL number No MDL found
  • Cas Number : 186203-81-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/11619.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/11619.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart