SY3428348195 Synthonix Corporation

Methyl 6-chloro-1,2,4-triazolo[4,3-b]pyridazine-3-carboxylate

  • MDL number No MDL found
  • Cas Number : 76196-04-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/10504.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/10504.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart