SY3428348211 Synthonix Corporation

1-Methyl-1,2,3,6-tetrahydropyridin-4-yl trifluoromethanesulfonate

  • MDL number No MDL found
  • Cas Number : 180692-27-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/10614.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/10614.html
hstatements H302 , H315 , H318 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart