SY3428348214 Synthonix Corporation

Methyl (2S,4S)-1-Boc-4-trifluoromethylpyrrolidine-2-carboxylate

  • MDL number No MDL found
  • Cas Number : 1523541-90-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/10655.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/10655.html
hstatements H302 , H315 , H318 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart