SY3428348237 Synthonix Corporation

Methyl 4-fluoro-1H-indazole-5-carboxylate

  • MDL number No MDL found
  • Cas Number : 473416-82-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/10763.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/10763.html
hstatements H302 , H315 , H318 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart