SY3428348359 Synthonix Corporation

(2S)-2-(Methoxymethyl)azetidine

  • MDL number MFCD11977228
  • Cas Number : 935668-27-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/11645.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/11645.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart