SY3428348549 Synthonix Corporation

Thiophene-3-ethylamine hydrochloride

  • MDL number No MDL found
  • Cas Number : 34843-84-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/10491.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/10491.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart