SY3432447541 Synthonix Corporation

4-Amino-1-Cbz-piperidine

  • MDL number No MDL found
  • Cas Number : 120278-07-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/12336.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/12336.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart