SY3432447552 Synthonix Corporation

1-Cbz-4-hydroxymethylpiperidine

  • MDL number No MDL found
  • Cas Number : 122860-33-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/12353.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/12353.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart