SY3432447593 Synthonix Corporation

3-Iodoimidazo[1,2-a]pyridine-6-carboxylic acid methyl ester

  • MDL number No MDL found
  • Cas Number : 460087-82-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/12394.html

Safety Information

MSDS https://synthonix.com/12394.html
hstatements H302 , H320 ,
Availability details for
Quantity
  • Availability
  • Add to Cart