SY3432447813 Synthonix Corporation

cis-1-Boc-3-hydroxy-4-aminomethylpyrrolidine

  • MDL number No MDL found
  • Cas Number : 872714-78-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/12624.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/12624.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart