SY3432447870 Synthonix Corporation

4-Bromo-1-benzothiophene-2-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 5194-37-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/12683.html

Safety Information

MSDS https://synthonix.com/12683.html
hstatements H302 , H320 ,
Availability details for
Quantity
  • Availability
  • Add to Cart