SY3432447888 Synthonix Corporation

2,4-Dichloro-6-methyl-nicotinic acid ethyl ester

  • MDL number MFCD00173918
  • Cas Number : 86129-63-7
  • Molecular Weight : 234.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/12701.html

Safety Information

signalword Warning
pcodes P202, P261, P280
MSDS https://synthonix.com/12701.html
pictograms GHS07
hstatements H302, H313, H315, H320
Availability details for
Quantity
  • Availability
  • Add to Cart