SY3432447986 Synthonix Corporation

3-Chloro-7-nitro-1H-indazole

  • MDL number No MDL found
  • Cas Number : 74209-33-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/12799.html

Safety Information

MSDS https://synthonix.com/12799.html
hstatements H302 , H313 , H315 , H320 ,
Availability details for
Quantity
  • Availability
  • Add to Cart