SY3432448042 Synthonix Corporation

3-Bromo-2-methoxy-4-picoline

  • MDL number No MDL found
  • Cas Number : 717843-51-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/12856.html

Safety Information

MSDS https://synthonix.com/12856.html
hstatements H302 , H320 ,
Availability details for
Quantity
  • Availability
  • Add to Cart