SY3432448056 Synthonix Corporation

Methyl 4-fluoro-1-(triisopropylsilanyl)-7-azaindole-5-carboxylate

  • MDL number MFCD11616413
  • Cas Number : 685513-92-2
  • Molecular Weight : 350.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 4-fluoro-1-(triisopropylsilyl)-1H-pyrrolo[2,3-b]pyridine-5-carboxylate
Inchi Code 1S/C18H27FN2O2Si/c1-11(2)24(12(3)4,13(5)6)21-9-8-14-16(19)15(18(22)23-7)10-20-17(14)21/h8-13H,1-7H3
Inchi Key XZCYLQFNGHAFPO-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Refrigerator
Physical Form oil
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/12870.html

Safety Information

signalword Warning
pcodes P202, P261, P280
MSDS https://synthonix.com/12870.html
pictograms GHS07
hstatements H320
Availability details for
Quantity
  • Availability
  • Add to Cart