SY3432448241 Synthonix Corporation

(Tetrahydro-2H-thiopyran-4-yl)methanamine

  • MDL number No MDL found
  • Cas Number : 666263-17-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/13090.html

Safety Information

MSDS https://synthonix.com/13090.html
hstatements H302 , H320 ,
Availability details for
Quantity
  • Availability
  • Add to Cart