SY3432448242 Synthonix Corporation

Tetrahydro-2H-thiopyran-4-carboxylic acid

  • MDL number MFCD09879921
  • Cas Number : 89489-53-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/13091.html

Safety Information

MSDS https://synthonix.com/13091.html
hstatements H302 , H320 ,
Availability details for
Quantity
  • Availability
  • Add to Cart