SY3432448358 Synthonix Corporation

Methyl exo-3-azabicyclo[3.1.0]hexane-6-carboxylate hydrochloride

  • MDL number No MDL found
  • Cas Number : 1212063-26-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/13240.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/13240.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart