SY3432448416 Synthonix Corporation

8-Iodo-3H-pyrazolo[1,5-a][1,3,5]triazin-4-one

  • MDL number No MDL found
  • Cas Number : 147916-84-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/13622.html

Safety Information

MSDS https://synthonix.com/13622.html
hstatements H302 , H320 ,
Availability details for
Quantity
  • Availability
  • Add to Cart