SY3432448599 Synthonix Corporation

Methyl (S)-1-Boc-3-fluoropyrrolidine-3-carboxylate

  • MDL number No MDL found
  • Cas Number : 1438852-71-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/13552.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/13552.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart