SY3432448751 Synthonix Corporation

3-Fluoro-4-nitroaniline

  • MDL number MFCD03412239
  • Cas Number : 2369-13-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Refrigerator
Physical Form Crystal - Powder
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/13749.html

Safety Information

signalword Warning
MSDS https://synthonix.com/13749.html
pictograms GHS07
hstatements H320
Availability details for
Quantity
  • Availability
  • Add to Cart