SY3432448775 Synthonix Corporation

Methyl 6-Cyanopyridine-3-carboxylate

  • MDL number No MDL found
  • Cas Number : 89809-65-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/13779.html

Safety Information

MSDS https://synthonix.com/13779.html
hstatements H302 , H320 ,
Availability details for
Quantity
  • Availability
  • Add to Cart