SY3432448790 Synthonix Corporation

5-Methyl-4,5,6,7-tetrahydro[1,3]thiazolo[5,4-c]pyridin-2-carboxylic acid hydrochloride

  • MDL number MFCD23106243
  • Cas Number : 1822641-87-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/13795.html

Safety Information

MSDS https://synthonix.com/13795.html
hstatements H302 , H320 ,
Availability details for
Quantity
  • Availability
  • Add to Cart