SY3432448815 Synthonix Corporation

7-(phenylsulfonyl)-7H-Pyrrolo[2,3-d]pyrimidine

  • MDL number No MDL found
  • Cas Number : 1279872-89-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/13854.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/13854.html
hstatements H302 , H315 , H319 , H320 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart